Mupirocin lithium
Bottle, 1 g
Katalognr. APOSBIL9900-1G
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
Synonyme: Lithium 9-(((E)-4-((2S,3R,4R,5S)-3,4-dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)nonanoate
- CAS Nummer:73346-79-9
- Formel:C₂₆H₄₃LiO₉
- MDL-Nummer:MFCD11977847
- Molekulargewicht:506.561 g/mol
- Lagertemperatur:Raumtemperatur
Technische Daten
Technische Daten
- Packung:Bottle
- VE:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



