Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Hydrogen Peroxide Blocking Reagent, Abcam

Hydrogen Peroxide Blocking Reagent, Abcam

Hydrogen Peroxide Blocking Reagent
Catalog # ABCAAB64218-60
Supplier:   Abcam
Hydrogen Peroxide Blocking Reagent, Abcam
Catalog # ABCAAB64218-60
Supplier:  Abcam
CAS Number:  
Hydrogen Peroxide Blocking Reagent, Abcam

Hydrogen Peroxide Blocking Reagent

CAS Number:  
Price
SFr. 130.00
Log in for availability
1 * 60 mL
Product Details & Documents

Hydrogen peroxide blocking reagent is used to block endogenous peroxidase activity in IHC applications, enhancing signal detection in HRP / peroxidase-based systems. - Cited in over 30 publications - Endogenous peroxidase block – prevents non-specific background staining in IHC - HRP-based detection – ideal for use with HRP/peroxidase detection systems - Concentration – contains 0.3% hydrogen peroxide

  • Preservative: 0.1% Sodium azide Constituents: 0.3% Hydrogen peroxide

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Bezeichnung:
    Hydrogen Peroxide Blocking Reagent
  • Cat. No.:
    ABCAAB64218-60
  • Lagertemperatur:
    Store at +4°C.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.