Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Human Recombinant IL-1 beta (from E. coli)

Human Recombinant IL-1 beta (from E. coli)

Katalognr. ABCAAB9617-2
Anbieter:   Abcam
Human Recombinant IL-1 beta (from E. coli)
Katalognr. ABCAAB9617-2
Anbieter:  Abcam
CAS Nummer:  
Human Recombinant IL-1 beta (from E. coli)
Human Recombinant IL-1 beta (from E. coli)
CAS Nummer:  
Preis
SFr. 165.00
Einloggen für Verfügbarkeit
1 * 2 µG
Product Details & Documents

Recombinant human IL-1 beta protein (Active) is a Human Full Length protein, in the 117 to 269 aa range, expressed in Escherichia coli, with >98% purity, <1 EU/µg endotoxin level and suitable for ELISA, FuncS, HPLC, SDS-PAGE.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Technische Daten

Technische Daten

  • Konjugation:
    Unconjugated
  • Form:
    Lyophilised
  • Transporttemperatur:
    20
  • Validierte Anwendungen:
    ELISA, FuncS, HPLC, SDS-PAGE
  • Purity:
    >98%
  • Spezies:
    Human
  • Biologische Aktivität:
    Bioactive
  • Zusammensetzung:
    Constituents: 0,3% Sodium chloride, 0,16% Sodium phosphate
  • Cat. No.:
    ABCAAB9617-2
  • Ursprung:
    E. coli
  • Endotoxingehalt:
    <1 EU/µg
  • Tierfrei:
    N
  • Protein/Peptid-Typ:
    Rekombinant
  • Lagerbedingungen:
    Store at –20 °C
  • Protein/Peptid-Name:
    IL-1 beta
  • Proteinsynonym:
    IL1F2|IL1B|Interleukin-1 beta|Catabolin

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.