AuftragsverfolgungKontakt
SureVector | Avantor
SureVector
SureVector Core kit, SureVector Core contains the basic elements needed for functionality in bacterial, yeast, and mammalian systems, 15 reactions, 1
Katalognr. AGLSG7514A
Anbieter:   AGILENT
SureVector
Katalognr. AGLSG7514A
Anbieter:  AGILENT
CAS Nummer:  
SureVector
SureVector
SureVector Core kit, SureVector Core contains the basic elements needed for functionality in bacterial, yeast, and mammalian systems, 15 reactions, 1
CAS Nummer:  
Preis
SFr. 802.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Combine standard DNA components to build custom vector constructs from Agilent’s set of standard parts.
  • Rapid custom vector generation - Less than a day from design to vector, compared to 4 weeks for custom vector services
  • Reliable and precise assembly - SureVector is extensively validated to ensure that standard parts can be interchanged, without loss of functionality
  • More flexible than traditional systems - Assemble new vectors in your lab as experimental requirements change, versus always ordering a new one
  • Control your experiments - Take control of your experiments, troubleshoot your own work, not that of your service providers DNA assembly
SureVector next-generation cloning uses the power of synthetic biology to combine standard DNA components to build custom vector constructs from the set of standard parts. Simply design a vector, order or PCR your gene-of-interest, and your custom plasmid is just a single reaction away.
Proprietary enzymes enable rapid and reliable assembly of multiple DNA fragments into a circular, recombinant plasmid containing your target gene.
Lieferinformationen: Components include: bacterial selection markers, bacterial origins of replication, mammalian selection markers, promoters for yeast, mammalian cells and bacteria, and a variety of solubility/affinity tags; Enabling functionality in a range of cells.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.
Technische Daten
Technische Daten
  • VE:
    1
  • Bezeichnung:
    SureVector Core kit, SureVector Core contains the basic elements needed for functionality in bacterial, yeast, and mammalian systems, 15 reactions