Multiple affinity removal spin cartridge reagent kit
Multiple affinity removal spin cartridge reagent kit, 1 kt
Catalog # AGLS5188-5254
Supplier: AGILENT
Multiple affinity removal spin cartridge reagent kit
Catalog # AGLS5188-5254
Supplier: AGILENT
CAS Number:
Jump to:
Product Details & Documents
The affinity removal spin cartridge reagent kit is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
Delivery information: Kit includes Buffer A, 1 bottle (1 L) Buffer B, 1 bottle (1 L), Spin filters, 2 packs, Spin concentrators, 5 KDa MWCo (4 ml) 1 pack, Luer-Lok adapter 1 pack, Luer-Lok syringes (5 ml) 1 pack, Microtubes, 6 packs, Caps and plugs 1 pack, PTFE Luer-Lok needles 1 pack.
Specifications
Specifications
- VE:1 kt
- Bezeichnung:Multiple affinity removal spin cartridge reagent kit
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.