AuftragsverfolgungKontakt
Multiple affinity removal spin cartridge reagent kit
Multiple affinity removal spin cartridge reagent kit
Multiple affinity removal spin cartridge reagent kit, 1 kt
Catalog # AGLS5188-5254
Supplier:   AGILENT
Multiple affinity removal spin cartridge reagent kit
Catalog # AGLS5188-5254
Supplier:  AGILENT
CAS Number:  
Multiple affinity removal spin cartridge reagent kit
Multiple affinity removal spin cartridge reagent kit, 1 kt
CAS Number:  
Price
SFr. 1’310.00
Log in for availability
1 * 1 kt
Product Details & Documents
The affinity removal spin cartridge reagent kit is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
Delivery information: Kit includes Buffer A, 1 bottle (1 L) Buffer B, 1 bottle (1 L), Spin filters, 2 packs, Spin concentrators, 5 KDa MWCo (4 ml) 1 pack, Luer-Lok adapter 1 pack, Luer-Lok syringes (5 ml) 1 pack, Microtubes, 6 packs, Caps and plugs 1 pack, PTFE Luer-Lok needles 1 pack.
Specifications
Specifications
  • VE:
    1 kt
  • Bezeichnung:
    Multiple affinity removal spin cartridge reagent kit