Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Multiple affinity removal spin cartridge reagent kit

Multiple affinity removal spin cartridge reagent kit

Microtube screwtop, 1,5 ml, 100
Catalog # AGLS5188-5251
Supplier:   AGILENT
Multiple affinity removal spin cartridge reagent kit
Catalog # AGLS5188-5251
Supplier:  AGILENT
CAS Number:  
Multiple affinity removal spin cartridge reagent kit

Microtube screwtop, 1,5 ml, 100

CAS Number:  
Price
68.00 SFr.
Log in for availability
1 * 100 ST
Product Details & Documents

The affinity removal spin cartridge reagent kit is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as plasma, serum, urine or cerebrospinal fluid).

Delivery information: Kit includes Buffer A, 1 bottle (1 L) Buffer B, 1 bottle (1 L), Spin filters, 2 packs, Spin concentrators, 5 KDa MWCo (4 ml) 1 pack, Luer-Lok adapter 1 pack, Luer-Lok syringes (5 ml) 1 pack, Microtubes, 6 packs, Caps and plugs 1 pack, PTFE Luer-Lok needles 1 pack.

Specifications

Specifications

  • VE:
    100
  • Bezeichnung:
    Microtube screwtop, 1,5 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.