Multiple affinity removal spin cartridge reagent kit
Syringes plastic Luer-lock, 5 ml, 2
Katalognr. AGLS5188-5250
Anbieter: AGILENT
Multiple affinity removal spin cartridge reagent kit
Katalognr. AGLS5188-5250
Anbieter: AGILENT
CAS Nummer:
Direkt zu
Product Details & Documents
The affinity removal spin cartridge reagent kit is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
: Kit includes Buffer A, 1 bottle (1 L) Buffer B, 1 bottle (1 L), Spin filters, 2 packs, Spin concentrators, 5 KDa MWCo (4 ml) 1 pack, Luer-Lok adapter 1 pack, Luer-Lok syringes (5 ml) 1 pack, Microtubes, 6 packs, Caps and plugs 1 pack, PTFE Luer-Lok needles 1 pack.
Technische Daten
Technische Daten
- VE:2
- Bezeichnung:Syringes plastic Luer-lock, 5 ml



