Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Dimetridazol 98%

Dimetridazol 98%

Bottle, 100 g
Catalog # APOSOR927600-100G
Supplier:   Apollo Scientific
Dimetridazol 98%
Catalog # APOSOR927600-100G
Supplier:  Apollo Scientific
CAS Number:  551-92-8
Dimetridazol 98%

Bottle, 100 g

CAS Number:  551-92-8
Price
SFr. 29.50
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,2-Dimethyl-5-nitroimidazole
  • CAS Number:551-92-8
  • Formula:C₅H₇N₃O₂
  • MDL Number:MFCD00047046
  • Molecular Weight:141.13 g/mol
  • Storage Temperature:Raumtemperatur
Specifications

Specifications

  • Packung:
    Bottle
  • VE:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.