4,4-Difluorocyclohexan-1-amine 97%
Bottle, 250 mg
Katalognr. APOSPC901499-250MG
Anbieter: Apollo Scientific
4,4-Difluorocyclohexan-1-amine 97%
Katalognr. APOSPC901499-250MG
Anbieter: Apollo Scientific
CAS Nummer: 458566-84-2
Direkt zu
Product Details & Documents
Synonyme: 4,4-Difluorocyclohexanamine
- CAS Nummer:458566-84-2
- Formel:C₆H₁₁F₂N
- MDL-Nummer:MFCD06409984
- Molekulargewicht:135.155 g/mol
- Lagertemperatur:Raumtemperatur
Technische Daten
Technische Daten
- Packung:Bottle
- VE:250 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



