Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
2-(Methylthio)benzaldehyd 90%

2-(Methylthio)benzaldehyd 90%

Bottle, 5 g
Catalog # APOSOR938789-5G
Irritant
Supplier:   Apollo Scientific
2-(Methylthio)benzaldehyd 90%
Catalog # APOSOR938789-5G
Supplier:  Apollo Scientific
CAS Number:  7022-45-9
2-(Methylthio)benzaldehyd 90%

Bottle, 5 g

CAS Number:  7022-45-9
Price
SFr. 148.00
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:7022-45-9
  • Formula:C₈H₈OS
  • MDL Number:MFCD00196822
  • Molecular Weight:152.2135 g/mol
  • Boiling Pt:141…143 °C
  • Density:1,18 g/cm³ (25 °C)
  • Storage Temperature:Raumtemperatur
Specifications

Specifications

  • Packung:
    Bottle
  • VE:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.