Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
4-Ethinylanisol 98%

4-Ethinylanisol 98%

Bottle, 100 g
Catalog # APOSOR21951-100G
Irritant
Supplier:   Apollo Scientific
4-Ethinylanisol 98%
Catalog # APOSOR21951-100G
Supplier:  Apollo Scientific
CAS Number:  768-60-5
4-Ethinylanisol 98%

Bottle, 100 g

CAS Number:  768-60-5
Price
SFr. 255.00
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (4-Methoxyphenyl)ethyne, (4-Methoxyphenyl)acetylene, 1-Ethynyl-4-methoxybenzene, 1-Methoxy-4-ethynylbenzene
  • CAS Number:768-60-5
  • Formula:C₉H₈O
  • MDL Number:MFCD00168815
  • Molecular Weight:132.16 g/mol
  • Boiling Pt:87…91 °C/11mmHg
  • Melting Pt:28…29 °C
  • Density:1,019 g/cm³ (25 °C)
  • Flash Pt:82 °C
  • Storage Temperature:Raumtemperatur

Documents

Specifications

Specifications

  • Packung:
    Bottle
  • VE:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.