4-Hydroxy-2,6-dimethoxybenzaldehyd 98%
Bottle, 5 g
Katalognr. APOSOR10495-5G
Anbieter: Apollo Scientific
4-Hydroxy-2,6-dimethoxybenzaldehyd 98%
Katalognr. APOSOR10495-5G
Anbieter: Apollo Scientific
CAS Nummer: 22080-96-2
Direkt zu
Product Details & Documents
Synonyme: 2,6-Dimethoxy-4-hydroxybenzaldehyde
- CAS Nummer:22080-96-2
- Formel:C₉H₁₀O₄
- MDL-Nummer:MFCD01407645
- Molekulargewicht:182.1733 g/mol
- Schmelzender Platingehalt:225 °C (Zers.)
- Schmelzender Platingehalt:225 °C (Zersetzung)
- Lagertemperatur:Raumtemperatur
Technische Daten
Technische Daten
- Packung:Bottle
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



