Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
4-Hydroxy-2,6-dimethoxybenzaldehyd 98%

4-Hydroxy-2,6-dimethoxybenzaldehyd 98%

Bottle, 25 g
Catalog # APOSOR10495-25G
Irritant
Supplier:   Apollo Scientific
4-Hydroxy-2,6-dimethoxybenzaldehyd 98%
Catalog # APOSOR10495-25G
Supplier:  Apollo Scientific
CAS Number:  22080-96-2
4-Hydroxy-2,6-dimethoxybenzaldehyd 98%

Bottle, 25 g

CAS Number:  22080-96-2
Price
394.00 SFr.
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2,6-Dimethoxy-4-hydroxybenzaldehyde
  • CAS Number:22080-96-2
  • Formula:C₉H₁₀O₄
  • MDL Number:MFCD01407645
  • Molecular Weight:182.1733 g/mol
  • Melting Pt:225 °C (Zers.)
  • Melting Pt:225 °C (Zersetzung)
  • Storage Temperature:Raumtemperatur
Specifications

Specifications

  • Packung:
    Bottle
  • VE:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.