(3-Fluorophenoxy)acetic acid
Bottle, 10 g
Katalognr. APOSPC200534-10G
Anbieter: Apollo Scientific
(3-Fluorophenoxy)acetic acid
Katalognr. APOSPC200534-10G
Anbieter: Apollo Scientific
CAS Nummer: 404-98-8
Direkt zu
Product Details & Documents
Synonyme: 2-(3-Fluorophenoxy)acetic acid
- CAS Nummer:404-98-8
- Formel:C₈H₇FO₃
- MDL-Nummer:MFCD02295721
- Molekulargewicht:170.14 g/mol
- Lagertemperatur:Raumtemperatur
Technische Daten
Technische Daten
- Packung:Bottle
- VE:10 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



