ContactUsLinkComponent
RAUSCHERT ultra membran Support: single channel tube da = 10 mm +0,6/-0,1 di = 7 mm +/-0,2 l = 600 mm, Al2O3 Membrane: ZrO2, d50 = 3 nm Endsealing: glass, approx. 13 mm on both ends 1 * 1 ST
Catalog # MSPPAA0600-A3Z3G
RAUSCHERT ultra membran Support: single channel tube da = 10 mm +0,6/-0,1 di = 7 mm +/-0,2 l = 600 mm, Al2O3 Membrane: ZrO2, d50 = 3 nm Endsealing: glass, approx. 13 mm on both ends 1 * 1 ST
Catalog # MSPPAA0600-A3Z3G
Frequently Bought Together