AccuScript high-fidelity reverse transcriptase
AccuScript reverse transcriptase, Engineered to deliver the highest transcription accuracy while promoting full-length cDNA synthesis, yield, and RT-PCR performance, 50 reactions, 1
Katalognr. AGLS600089
Anbieter: AGILENT
Direkt zu
Product Details & Documents
AccuScript High-Fidelity Reverse Transcriptase delivers the highest RT accuracy currently available.
- MMLV-RT derivative combined with 3 to 5’ exonuclease reduces errors
- High yields of full length cDNA up to 20 kb;RT-PCR kit synthesises cDNA from 10 to 1000 ng of total RNA or 0,1 to 10 ng of poly(A)+ RNA
- Stringent quality control procedures ensure high yields of premium cDNA
AccuScript High-Fidelity Reverse Transcriptase (RT) delivers 3,7 times more accuracy than other commercially available RTs and 6,6 times more accuracy than AMV RT. The AccuScript PfuUltra II RT-PCR kit provides a complete system in applications that demand high fidelity, specificity, and throughput.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.
Technische Daten
Technische Daten
- VE:1
- Bezeichnung:AccuScript reverse transcriptase
- Details:Engineered to deliver the highest transcription accuracy while promoting full-length cDNA synthesis, yield, and RT-PCR performance
- No. of reactions:50 reactions
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.