Universal mat rolls, PIG®
Product Details & Documents
Eight layers of 100% polypropylene are thermally bonded to make PIG® Mat strong; won't rip, tear or fray even when saturated.
- Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
- Highly absorbent, fine-fibre construction won't leave behind liquids or fibre residue
- Light-weight construction is ideal for small tasks and clean-ups
- Perforated every 25 cm, so you only use what you need
- Smaller roll length for counter display; packaged ready to sell straight to customers
- Can be used with a 12-roll point-of-sale display box for counter promotion
- Absorbs oils, coolants, solvents, and water
Perfectly sized mat rolls thoroughly outperform wipes, rags and loose absorbents for a cleaner shop environment. Exceptional durability - eight layers of 100% polypropylene make PIG® Mat the strongest mat on the market, It won't rip, tear or fray even when saturated. Less waste - with PIG® Mat, you'll absorb way more liquid with less so no need to go through a big handful of wipes for every small job.
Super wicking powers - exclusive dimple pattern speeds wicking of liquid throughout the mat for faster, easier cleanup. Versatility - PIG® Mat can do anything wipes and rags can do and then some line toolboxes, benches and floors and clean up spills in addition to drying and wiping tasks. Convenience - It's easy to find a place for these compact, grab-and-go rolls throughout the shop. Add a magnetic roll holder to attach one to your toolbox or maintenance cart.
Technische Daten
- Bezeichnung:Universal mat roll, German label, 1,2 kg, 100% polypropylene
- L×B:15 m × 380 mm
- Seq:4
- Absorptionsvermögen:12,5 L
- Verpackt:1 roll
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



