Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Tyloxapol USP

Tyloxapol USP

Glass, amber, 100 g
Codice catalogo SPCMT1708-100GMBL
Nome del fornitore:   Spectrum Chemical
Tyloxapol USP
Codice catalogo SPCMT1708-100GMBL
Nome del fornitore:  Spectrum Chemical
Numero CAS:  25301-02-4
Tyloxapol USP

Glass, amber, 100 g

Numero CAS:  25301-02-4
Price
SFr. 2’660.00
Log in per disponibilità
1 * 100 g
Product Details & Documents

All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

  • Numero CAS:25301-02-4
  • Numero MDL:MFCD00149002
  • Densità:1,100
  • temperatura di conservazione:Raumtemperatur
Specifiche

Specifiche

  • Packung:
    Glass, amber
  • VE:
    100 g

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.