Fraction collectors
Product Details & Documents
Ideal solutions for small to large capacity collection. Gilson offers advanced solutions for fraction collection with a complete line of stand‑alone and software-controlled fraction collectors ranging from small footprint to large vessel capacity models. The built-in, easy-to-use keypad makes setup and operation simple and capable of interfacing with any HPLC system or control the system through your PC with TRILUTION® LC software.
- Collect fractions by time, drop, or peak to suit all your LC applications
- Choose the compact FC 203B for small-capacity collection or the FC 204 for large volumes and capacities
- In peak mode, collectors monitor a detector signal and identify peaks using either an adaptive-slope algorithm or a specified millivolt threshold
- Time: Up to 10 programmable time windows can be added for collecting only what you want, while discarding the column’s void volume, peaks of no interest, and equilibration volumes
- 3-way diverter valve prevents contamination of collected fractions (optional)
- Compact size fits in most fume hoods
FC203B and FC203B with valve: choose among peak, drop, or time collection options with the FC 203B fraction collector. In peak mode, the FC 203B monitors a detector signal and identifies peaks using either an adaptive-slope algorithm or a specified millivolt threshold. With up to ten programmable time windows to use, collect only what you want and discard the column void volume, peaks of no interest, and equilibration volumes. Multicolumn adapters are available for collecting up to 18 separate channels and an optional 3-way diverter valve prevents contamination of collected fractions.
FC204: Collect samples by peak, drop, or time using the FC 204 Fraction Collector. Add up to ten programmable time windows to collect only what you want, eliminating void volumes, equilibration volumes, and peaks of no interest. In peak mode, the collector monitors a detector signal, identifying peaks using an adaptive-slope algorithm or a specified millivolt threshold. The 3-way diverter valve prevents contamination of fractions. Multicolumn adapters allow for collection from up to 18 separate channels.
Technische Daten
- Bezeichnung:Fraction collector FC204 with low pressure valve, HPLC purification technique, 110 - 120/220 - 240 V
- Seq:03
Accessories
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI150427Rack, code 24, holds quantity of 14, 28×57 mm scintilation vials (20 ml)Einloggen für Verfügbarkeit1 * 1 ST - SFr. 92.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI150420Rack, code 28, holds quantity of 108 of 10×75 mm tubes (3,5 ml)Einloggen für Verfügbarkeit1 * 1 ST - SFr. 91.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI150422Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml)Einloggen für Verfügbarkeit1 * 1 ST - SFr. 103.40
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI150424Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml), for use with FC204Einloggen für Verfügbarkeit1 * 1 ST - SFr. 200.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI150425Rack, code 20, holds quantity of 108, 10×100 mm tubes (4,5 ml)Einloggen für Verfügbarkeit1 * 1 ST - SFr. 142.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI150426Rack, code 23, holds quantity of 44, 17×55 mm scintilation vials (6,8 ml)Einloggen für Verfügbarkeit1 * 1 ST - SFr. 92.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI150429Rack, code 29, holds quantity of 60, 12/13×75 mm tubes ( 5/6 ml)Einloggen für Verfügbarkeit1 * 1 ST - SFr. 100.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI150498Rack, code 22U, Universal, holds 10 - 18×100 - 175 mm tubes (4 ml), for use with FC204Einloggen für Verfügbarkeit1 * 1 ST - SFr. 357.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI152420Rack, code 28, holds quantity of 108, 10×75 mm tubes (5/pack)Einloggen für Verfügbarkeit1 * 5 ST - SFr. 405.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI152422Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml) (5/pack)Einloggen für Verfügbarkeit1 * 5 ST - SFr. 357.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI152424Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml) (5/pack), for use with FC204Einloggen für Verfügbarkeit1 * 5 ST - SFr. 964.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI152426Rack, code 23, holds quantity of 44, 17×55 scintilation vials (5/pack)Einloggen für Verfügbarkeit1 * 5 ST - SFr. 430.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI152427Rack, code 24, holds quantity of 14, 28×57 mm scintilation vials (5/pack)Einloggen für Verfügbarkeit1 * 5 ST - SFr. 426.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI270433Rack, code 23W, holds quantity of 44, 15×45 mm WISP vials (4 ml)Einloggen für Verfügbarkeit1 * 1 ST - SFr. 282.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI170413Rack, code 11, holds quantity of 105, 13×100 mm, 9 ml, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 1’230.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI2704342Rack, code 29LE, holds quantity of 60, 1,5 ml microcentrifuge tubesEinloggen für Verfügbarkeit1 * 1 ST - SFr. 351.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI2704341Rack, code 29SE, holds quantity of 60, 0,5 ml microcentrifuge tubesEinloggen für Verfügbarkeit1 * 1 ST - SFr. 428.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI130412Rack, code 4, holds quantity of 80, 12×75 mm tubes, 5 ml, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 681.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI170414Rack, code 14, holds quantity of 128, 12×75 or 100 mm, 5 ml, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 970.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI270430Rack, code 0, holds quantity of 80, 12×32 mm vials, 2 ml, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 65.40
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI1707303-way valve kit with 1,5 mm Int.Ø, FC203, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 898.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI170735FC-204 Prep valve assembly package, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 1’630.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI170415Rack, code 15, holds 1×96-well microplate, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 114.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI170416Rack, code 16, holds 4×96-well microplates, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 241.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI170417Rack heightener, code 17, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 221.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI170418Rack, code 17, microplate, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 424.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI1707343-way valve kit with 1,5 mm Int.Ø, FC204, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 999.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI170416DRack, code 16D, holds 4 deep well microplates, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 241.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- GILI174732Multiple column adapter, for use with fraction collectorsEinloggen für Verfügbarkeit1 * 1 ST - SFr. 885.00
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.