Lead AAS standards, (CRMs), Fluka™
Blei, Pb, 10000 mg/L, 5% HNO₃, Plastic bottle, 100 mL
Katalognr. HONCE4115-100ML
Anbieter: Honeywell Chemicals
Lead AAS standards, (CRMs), Fluka™
Katalognr. HONCE4115-100ML
Anbieter: Honeywell Chemicals
CAS Nummer:
Direkt zu
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Technische Daten
Technische Daten
- Konzentration:10000 mg/L
- Matrix:5% HNO₃
- Packung:Plastic bottle
- VE:100 mL
- Element:Blei
- Symbol:Pb
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



