Lead AAS standards, (CRMs), Fluka™
Blei, Pb, 1000 mg/L, 2% HNO₃, Plastic bottle, 500 mL
Katalognr. HONCE4055-500ML
Anbieter: Honeywell Chemicals
Lead AAS standards, (CRMs), Fluka™
Katalognr. HONCE4055-500ML
Anbieter: Honeywell Chemicals
CAS Nummer:
Direkt zu
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Zertifizierung: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Technische Daten
Technische Daten
- Konzentration:1000 mg/L
- Matrix:2% HNO₃
- Packung:Plastic bottle
- VE:500 mL
- Element:Blei
- Symbol:Pb
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



