ICP standard solutions, Fluka™
ICP Standard (7 Components), 50 - 1000 mg/L, 5% HNO₃/0,2% HF, 100 mL
Katalognr. HONCE2217-100ML
Anbieter: Honeywell Chemicals
Direkt zu
Product Details & Documents
Certified Reference Materials (CRMs).
- Traceable to NIST
: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB)
Technische Daten
Technische Daten
- Konzentration:50 - 1000 mg/L
- Matrix:5% HNO₃/0,2% HF
- VE:100 mL
- Bezeichnung:ICP Standard (7 Components)



