ICP standard solutions, Fluka™
ICP Standard (4 Components), 1000 mg/L, 2% HNO₃, 100 mL
Katalognr. HONCE2216-100ML
Anbieter: Honeywell Chemicals
Direkt zu
Product Details & Documents
Certified Reference Materials (CRMs).
- Traceable to NIST
: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB)
Technische Daten
Technische Daten
- Konzentration:1000 mg/L
- Matrix:2% HNO₃
- VE:100 mL
- Bezeichnung:ICP Standard (4 Components)



