ContactUsLinkComponent
Custom-BG-VAR Custom TEM Grid mix - with 4 Plus, 4 Minus, and 4 Protein A SMART Grids, Si grids with 25 nm SiO2 membranes, 50x50 mic ron windows. (12 grids total) 1 * 1 ST
Katalognr. MSPPCUSTOM-BG-VAR
Custom-BG-VAR Custom TEM Grid mix - with 4 Plus, 4 Minus, and 4 Protein A SMART Grids, Si grids with 25 nm SiO2 membranes, 50x50 mic ron windows. (12 grids total) 1 * 1 ST
Katalognr. MSPPCUSTOM-BG-VAR
Frequently Bought Together