ContactUsLinkComponent
BENZOIC ACID CALIBRANT FOR QNMR 5mM BENZOIC ACID IN DMSO-D6 (750UL FILL VOLUME) 1 * 1 ST
Katalognr. CAMBDLM-99491C
BENZOIC ACID CALIBRANT FOR QNMR 5mM BENZOIC ACID IN DMSO-D6 (750UL FILL VOLUME) 1 * 1 ST
Katalognr. CAMBDLM-99491C
Frequently Bought Together