AuftragsverfolgungKontakt
Storage Cleaners Cabinets | Avantor
Storage Cleaners Cabinets
Cleaners cabinet on leveler legs, 2, Edelstahl 304, 500×800×1800 mm
Katalognr. PALBJC0850
Anbieter:   Palbam
Storage Cleaners Cabinets
Katalognr. PALBJC0850
Anbieter:  Palbam
Storage Cleaners Cabinets
Cleaners cabinet on leveler legs, 2, Edelstahl 304, 500×800×1800 mm
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Designed for storing mops and cleaning materials.
  • Two doors
Fabrication material stainless steel 304 with electropolished finish.
Technische Daten
  • Bezeichnung:
    Cleaners cabinet on leveler legs
  • Material:
    Edelstahl 304
  • Seq:
    02
  • B×T×H:
    500×800×1800 mm
  • Türen:
    2
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..