(±)-Epichlorhydrin, GPR RECTAPUR®
1 L
: (±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane
- :106-89-8
- :C₃H₅ClO
- :MFCD00005132
- :92.52 g/mol
- :13,03642
- :117,9 °C (1013 hPa)
- :–57,2 °C
- :0,9985 g/cm³ (20 °C)
- :28 °C
- :Raumtemperatur
- :203-439-8
- :2023
- :6.1 (3),II
- Appearance :Clear liquid
- Assay (GC) :≥99.0 %
- Density (d 20 °C/ 4 °C) :1.178 - 1.182
- Identity (IR) :passes test
- VE:1 L



