CoolRack XT PCR-M for 12 x 1.5/2 ml tubes and 6 x PCR strips compatible with CoolBox XT, CoolBox 2XT 1 * 1 ST
Catalog # BZYM210184
Supplier: BIOZYM
CoolRack XT PCR-M for 12 x 1.5/2 ml tubes and 6 x PCR strips compatible with CoolBox XT, CoolBox 2XT 1 * 1 ST
Catalog # BZYM210184
Supplier: BIOZYM
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Serologische Pipetten, Standard Serie
Supplier: VWR Collection
Nitrilhandschuhe, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Nitrilhandschuhe
Supplier: VWR Collection
VWR® Gesichtstücher
Supplier: VWR Collection
Acetonitril ≥99,9%, HiPerSolv CHROMANORM® für die LC-MS, geeignet für UPLC/UHPLC-Instrumente
Supplier: VWR Chemicals
pH-Indikatorstäbchen, MQuant®, Supelco®
Supplier: Merck
Wasser, HiPerSolv CHROMANORM® für die HPLC
Supplier: VWR Chemicals
Methanol ≥99,8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Gradienten-Qualität für die HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrilhandschuhe
Supplier: VWR Collection
VWR®, Transferpipetten, High Performance
Supplier: VWR Collection