ContactUsLinkComponent
Human PDGF-BB, Tag Free (PDB-H4112) is expressed from E.coli cells. It contains AA Ser 82 - Thr 190 (Accession P01127-1). 1 * 20 µG
Katalognr. ACRBPDB-H4112-20UG
Anbieter:  ACRO Biosystems
Human PDGF-BB, Tag Free (PDB-H4112) is expressed from E.coli cells. It contains AA Ser 82 - Thr 190 (Accession P01127-1). 1 * 20 µG
Katalognr. ACRBPDB-H4112-20UG
Anbieter:  ACRO Biosystems
Frequently Bought Together