Gum tragacanth, Pulver NF
Poly bottle, 500 g
Anbieter: Spectrum Chemical
Gum tragacanth, Pulver NF
Katalognr. SPCMTR105-500GM
Anbieter: Spectrum Chemical
CAS Nummer: 9000-65-1
Direkt zu
Product Details & Documents
Tragacanth, Powder, NF is used as an emulsifier, thickener, stabiliser, and texturant additive.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Synonyme: Tragacanth, Tragacanth gum
- CAS Nummer:9000-65-1
- MDL-Nummer:MFCD00131255
- Lagertemperatur:Raumtemperatur
- EINECS:232-552-5
Technische Daten
Technische Daten
- Packung:Poly bottle
- VE:500 g



