Gum tragacanth, Pulver NF
Poly bottle, 125 g
Katalognr. SPCMTR105-125GM
Anbieter: Spectrum Chemical
Gum tragacanth, Pulver NF
Katalognr. SPCMTR105-125GM
Anbieter: Spectrum Chemical
CAS Nummer: 9000-65-1
Direkt zu
Product Details & Documents
Tragacanth, Powder, NF is used as an emulsifier, thickener, stabiliser, and texturant additive.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Synonyme: Tragacanth, Tragacanth gum
- CAS Nummer:9000-65-1
- MDL-Nummer:MFCD00131255
- Lagertemperatur:Raumtemperatur
- EINECS:232-552-5
Technische Daten
Technische Daten
- Packung:Poly bottle
- VE:125 g



