Gum tragacanth, Pulver NF
Poly pail, 12 kg
Katalognr. SPCMTR105-12KG
Anbieter: Spectrum Chemical
Gum tragacanth, Pulver NF
Katalognr. SPCMTR105-12KG
Anbieter: Spectrum Chemical
CAS Nummer: 9000-65-1
Direkt zu
Product Details & Documents
Tragacanth, Powder, NF is used as an emulsifier, thickener, stabiliser, and texturant additive.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Synonyme: Tragacanth, Tragacanth gum
- CAS Nummer:9000-65-1
- MDL-Nummer:MFCD00131255
- Lagertemperatur:Raumtemperatur
- EINECS:232-552-5
Documents
Technische Daten
Technische Daten
- Packung:Poly pail
- VE:12 kg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



