Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Quantitative filter papers, pyramid folded, Whatman™

Quantitative filter papers, pyramid folded, Whatman™

4, 125 mm
Katalognr. WHAT989810112
Quantitative filter papers, pyramid folded, Whatman™
Katalognr. WHAT989810112
Quantitative filter papers, pyramid folded, Whatman™
Quantitative filter papers, pyramid folded, Whatman™

4, 125 mm

Preis
SFr. 813.00
Einloggen für Verfügbarkeit
1 * 1.000 ST
Product Details & Documents

Easily fits into conical funnels.

  • High quality and consistency
  • Easy to use
Technische Daten
  • Sorte:
    4
  • Seq:
    04
  • Größe:
    125 mm
  • Cat. No.:
    WHAT989810112
  • Aschegehalt:
    0,007%
  • Stärke:
    215 µm
  • Gewicht:
    85 g/m²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.