Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Chitosan, Pulver | Avantor

Chitosan, Pulver

1 kg
Katalognr. SPCMC1569-1KGBL
Anbieter:   Spectrum Chemical
Chitosan, Pulver
Katalognr. SPCMC1569-1KGBL
Anbieter:  Spectrum Chemical
CAS Nummer:  9012-76-4
Chitosan, Pulver

1 kg

CAS Nummer:  9012-76-4
Preis
SFr. 938.00
Einloggen für Verfügbarkeit
1 * 1 kg
Product Details & Documents

Chitosan, Powder is a polysaccharide made by treating shrimp and other crustacean shells with the alkali sodium hydroxide. It can be used in agriculture as a seed treatment and to fight off fungal infections and is often used in the wine making industry as a chemical to prevent spoilage. In medicine it can be used in bandages to reduce bleeding and as an antibacterial agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chitosan can be used in agriculture as a seed treatment and biopesticide to fight off fungal infections and is often used in the wine making industry as a chemical to prevent spoilage. In medicine it can be used in bandages to reduce bleeding and as an antibacterial agent.

Synonyme:  Deacetylated chitin, Poly(D-glucosamine)
  • CAS Nummer:9012-76-4
  • MDL-Nummer:MFCD00161512
  • Lagertemperatur:Raumtemperatur
Technische Daten

Ergebnisse des Spezifikationstests

  • Synonyms:
    β-(1,4)-2-Amino-2-deoxy-D-glucose; Deacetylated Chitin; Poly(D-glucosamine)
  • Form:
    Powder
  • Deacetylation:
    Min. 90.0 %
  • Moisture:
    Max. 10.0 %
  • Ash:
    Max. 5.0 %

Technische Daten

  • VE:
    1 kg

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.