Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Stereo microscopes, ergonomic, Mantis
Stereo microscopes, ergonomic, Mantis
MEO objective 10× for Mantis Elite
Katalognr. VISIMEO-010
Anbieter:   VISION ENGINEERING
Stereo microscopes, ergonomic, Mantis
Katalognr. VISIMEO-010
Anbieter:  VISION ENGINEERING
Stereo microscopes, ergonomic, Mantis
Stereo microscopes, ergonomic, Mantis
MEO objective 10× for Mantis Elite
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Superior imaging for a wide range of inspection and rework tasks.
  • Patented optical technology for fatigue-free viewing and superb image quality
  • Wide range of magnification options to 20x
  • Long working distances; large depths of field
  • Shadow-free true colour LED illumination
  • Choice of table stand or articulated arm
Mantis Compact excels in the low magnification range for inspection or manipulation tasks where bench magnifiers have traditionally been used. Mantis Compact has a small footprint and low nvestment cost giving a superb price/performance ratio.
Mantis Elite has enhanced optical performance, including higher magnification, a large field of view plus long working distances, making it a perfect alternative to traditional stereo microscopes for a wide range of inspection, preparation and manipulation tasks requiring hand-eye co-ordination.
Ordering information: Objective lenses sold separately.
Documents
Technische Daten
  • Bezeichnung:
    MEO objective 10× for Mantis Elite
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..