Safety spectacles, Rigi™
Rigi™ safety spectacles, Clear, anti-mist
Katalognr. JSPA1RIG23CKN
Anbieter: JSP
Direkt zu
Product Details & Documents
Ultra-lightweight safety spectacle with a wide field of vision to provide optimal coverage.
- Large field of vsion
- Wide range of lenses available
- Optimal coverage
Whatever the workplace environment, Rigi™ spectacles deliver the perfect combination of performance, safety, and active style. The unique close fitting design, and ultra light weight (19g) ensure lasting comfort and fit. With a wide field of vision and a design to fit all faces, wearers can achieve a comfortable customised fit throughout the day. The ergonomic fit with flat and thin flexofit temples, is compatible with any ear defender or helmet.
Zertifizierung: Meets EN 166:2001.
Bestellinformationen: Available in a wide range of lens versions, including excellence coating and a unique selection of Blue Blocker lenses.
Technische Daten
- Bezeichnung:Rigi™ safety spectacles
- Seq:02
- Verglasung:Clear, anti-mist