ContactUsLinkComponent
[EN]EPA 8260 INERNAL STD MIX 2000µg/ML 1 1 * 5 ST
Katalognr. EHERYS09000011ME
Anbieter:  EHRENSTORFER
[EN]EPA 8260 INERNAL STD MIX 2000µg/ML 1 1 * 5 ST
Katalognr. EHERYS09000011ME
Anbieter:  EHRENSTORFER
Frequently Bought Together