Natriumhydroxid 1 M für HPCE, Supelco®
100 mL
Katalognr. 79724-100ML
Anbieter: Merck
Direkt zu
Product Details & Documents
- Useful for transesterification of triglycerides, cholesteryl esters and phospholipids
- Provides convenient, fast, quantitative derivatisation
- Converts free fatty acids into its sodium salts
Synonyme: Caustic soda
- CAS Nummer:1310-73-2
- Formel:NaOH
- MDL-Nummer:MFCD00003548
- Molekulargewicht:40 g/mol
- UN:1824
- ADR:8,II
Documents
Zertifikate
Suche nach ChargennummerTechnische Daten
Ergebnisse des Spezifikationstests
- Appearance (Color):Colorless
- Appearance (form):Liquid
- Titration (Molarity):0.9 - 1.1 mol/l
- UV-Absorption Values:260nm ≤ 0.5, 270nm ≤ 0.2
- Test by UV Spectroscopy:280nm ≤ 0.03, 290nm ≤ 0.01, 500nm ≤ 0.01
- Luminescence:Corresponds to Requirements
- Residue (Filter Test):No Residue
- Filtertest (Method):Neat
- Recommended Retest Period:84 Months
Technische Daten
- VE:100 mL
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



