Biological indicators, self-contained tests, STERILtest
Medium for the germination of the Geobacillus stearothermophilus spores
Catalog # 20199-VWR
Supplier: LIOFILCHEM
Jump to:
Product Details & Documents
Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).
Devices with nutrient medium, pH indicator and discs with spores of Geobacillus stearothermophilus (GST) for steam sterilisation or Bacillus atropheus (BAT) for ethylene oxide sterilisation.
Specifications
- Bezeichnung:Medium for the germination of the Geobacillus stearothermophilus spores
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.