AuftragsverfolgungKontakt
Biological indicators, self-contained tests, STERILtest
Biological indicators, self-contained tests, STERILtest
Medium for the germination of the Geobacillus stearothermophilus spores
Catalog # 20199-VWR
Supplier:   LIOFILCHEM
Biological indicators, self-contained tests, STERILtest
Catalog # 20199-VWR
Supplier:  LIOFILCHEM
Biological indicators, self-contained tests, STERILtest
Biological indicators, self-contained tests, STERILtest
Medium for the germination of the Geobacillus stearothermophilus spores
Price
SFr. 74.90
Log in for availability
1 * 20 Tests
Product Details & Documents
Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).
Devices with nutrient medium, pH indicator and discs with spores of Geobacillus stearothermophilus (GST) for steam sterilisation or Bacillus atropheus (BAT) for ethylene oxide sterilisation.
Specifications
  • Bezeichnung:
    Medium for the germination of the Geobacillus stearothermophilus spores