Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Nicotine solution, NMR reference standard, Supelco®

Nicotine solution, NMR reference standard, Supelco®

Nicotine solution, 2 mg/mL in DMSO-d6 (99.9 atom % D), TMS 0.05 %, NMR reference standard, NMR tube size 5 mm x 8 in., 10
Catalog # 612030-10X8IN
Supplier:   Merck
Nicotine solution, NMR reference standard, Supelco®
Catalog # 612030-10X8IN
Supplier:  Merck
CAS Number:  
Nicotine solution, NMR reference standard, Supelco®

Nicotine solution, 2 mg/mL in DMSO-d6 (99.9 atom % D), TMS 0.05 %, NMR reference standard, NMR tube size 5 mm x 8 in., 10

CAS Number:  
Price
2 510.00 SFr.
Log in for availability
1 * 10 ST
Product Details & Documents
Specifications

Specifications

  • Konzentration:
    2 mg/mL in DMSO-d6 (99.9 atom % D), TMS 0.05 %
  • VE:
    10
  • Normen:
    NMR reference standard
  • Bezeichnung:
    Nicotine solution
  • Pk Info:
    NMR tube size 5 mm x 8 in.
  • Cat. No.:
    612030-10X8IN
  • Bezeichnung:
    Nicotine solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.