Warning signs
Warning of asphyxiating atmosphere, Polyester, 200×173 mm
Catalog # BRDY196001
Supplier: Brady
Jump to:
Product Details & Documents
These safety signs are for the purposes of accident prevention, fire protection, health hazard information and emergency evacuation.
- Recommended by ISO 7010
- Colour: Yellow/Black
- Shape: Triangle
Specifications
- Bezeichnung:Warning of asphyxiating atmosphere
- Abmessungen:200×173 mm
- Material:Polyester
- Seq:20