Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Broad range RNA quantitation kits, AccuBlue®

AccuBlue® broad range RNA quantitation kit, 1.000 Assays
Katalognr. 31073.
Anbieter:   Biotium
Broad range RNA quantitation kits, AccuBlue®
Katalognr. 31073.
Anbieter:  Biotium
CAS Nummer:  
Broad range RNA quantitation kits, AccuBlue®

AccuBlue® broad range RNA quantitation kit, 1.000 Assays

CAS Nummer:  
Preis
SFr. 605.00
Einloggen für Verfügbarkeit
1 * 1.000 Essai
Product Details & Documents

The AccuBlue® broad range RNA quantitation kit boasts the widest linear range (5 to 1000 ng RNA) of available RNA quantitation kits, while providing exceptional accuracy, sensitivity, and high RNA selectivity.

  • Detection range spans Qubit® HS and BR
  • Highly selective for RNA over dsDNA
  • Far-red fluorescence emission
  • For fluorescence microplate reader or Qubit® RNA broad range program
  • Mammalian RNA standard to accurately quantify similar samples

High sensitivity and broad linear range:
The assay is linear between 5 and 1000 ng of RNA per 200 μl assay well, which spans both Qubit® high sensitivity and Qubit® broad range detection ranges. It is ideal for use in quantifying RNA for sensitive applications such as Next-Gen Sequencing (NGS) or reverse transcription PCR (RT-PCR).

RNA specificity:
Unlike absorbance-based measurements, RNA broad range dye is selective for RNA over double-stranded DNA, and can tolerate an equimolar amount of dsDNA in the sample without significant effect on RNA quantitation (purified RNA samples are still recommended).

Instrument compatibility:
The AccuBlue® broad range RNA quantitation assay is designed for use with fluorescence microplate readers equipped with excitation and emission filters for detecting far-red fluorescence. It is also optimised for use in the Qubit® fluorometer, using the pre-programmed RNA Broad Range program.

 : Replacement vials of RNA standard can be purchased separately.

 : Kit components: RNA broad range dye 200X, RNA broad range buffer, RNA dilution buffer (Standard 1 if using Qubit®), RNA broad range standard, 100 ng/μl (Standard 2 if using Qubit®).

Technische Daten

Technische Daten

  • VE:
    1.000 Assays
  • Bezeichnung:
    AccuBlue® broad range RNA quantitation kit

Technische Daten