ContactUsLinkComponent
With screw cap made of PP. GL 63, Durchmesser 91mm, Höhe 226mm. Good chemical resistance, ideal for long-term storage of liquids. Autoclavable at 121 °C (2 bar) according to DIN EN 285. Wider mouth for simple filling, also ideal for powders and paste 1 * 12 ST
Katalognr. VITL94194
Anbieter:  VITLAB
With screw cap made of PP. GL 63, Durchmesser 91mm, Höhe 226mm. Good chemical resistance, ideal for long-term storage of liquids. Autoclavable at 121 °C (2 bar) according to DIN EN 285. Wider mouth for simple filling, also ideal for powders and paste 1 * 12 ST
Katalognr. VITL94194
Anbieter:  VITLAB
Frequently Bought Together