ContactUsLinkComponent
Zentrifugenglas, gerader Rand, 80 ml 80 ml, Rohr 38 x 2,8, Länge 115 mm, Boro 3.3 1 * 1 ST
Catalog # NEUB0441-080-38
Zentrifugenglas, gerader Rand, 80 ml 80 ml, Rohr 38 x 2,8, Länge 115 mm, Boro 3.3 1 * 1 ST
Catalog # NEUB0441-080-38
Frequently Bought Together