Thermal transfer printer labels, type B-8423
Printer labels, B-8423, BPT-G3-8423-YL, Polyester, Permanent, Acryl, Thermal transfer, Gelb, BBP11, 15×6 mm, 2500 labels/box
BRDY217053
: Brady
Printable satin polyester labels for thermal transfer printers.
- Die-cut labels are supplied on a roll
- Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
- Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL and CSA approved with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.
- Druckermodell:BBP11
- Printing method:Thermal transfer
- Typ:B-8423
- Farbe:Gelb
- Bezeichnung:Printer labels
- Abmessungen:15×6 mm
- Material:Polyester
- Seq:02
- Modell:BPT-G3-8423-YL
- Verpackt:2500 labels/box
- Klebstoff:Permanent, Acryl



