Echinocandin B
5 mg
Katalognr. CAYM25520-5
Anbieter: Cayman Chemical
Direkt zu
Product Details & Documents
Echinocandin B inhibits 1,3-β glucan synthase; inhibits incorporation of radiolabeled glucose into alkali-insoluble glucan by S. cerevisiae spheroplasts; inhibits growth of C. albicans in vitro (MIC = 3.9 µg/ml after 2 days)
- An echinocandin antifungal
- Stability: ≥2 years
Dokumente
CAYM25520-5_TDS_EN
Technisches Datenblatt
Echinocandin B
Weitere Produktdetails
Technische Daten
Technische Daten
- VE:5 mg
- Cat. No.:CAYM25520-5
- CAS Nr.:54651-05-7
- Form:Powder
- Purity:>95%
- Lagerbedingungen:Freezer
- Protein/Peptid-Name:Echinocandin B
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



