Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Warning signs
Warning signs
Warning of explosive atmosphere, Polyester, 87×100 mm
Catalog # BRDY810882
Supplier:   Brady
Warning signs
Catalog # BRDY810882
Supplier:  Brady
Warning signs
Warning signs
Warning of explosive atmosphere, Polyester, 87×100 mm
Price
13.30 SFr.
Log in for availability
1 * 3 ST
Product Details & Documents
These safety signs are for the purposes of accident prevention, fire protection, health hazard information and emergency evacuation.
  • Recommended by ISO 7010
  • Colour: Yellow/Black
  • Shape: Triangle
Specifications
  • Bezeichnung:
    Warning of explosive atmosphere
  • Abmessungen:
    87×100 mm
  • Material:
    Polyester
  • Seq:
    13