Guard chips, Intuvo
Product Details & Documents
Features and specifications for the Intuvo Guard Chips.
- Saves time and improves sample throughput; no more column trimming
- Enables retention time reproducibility, and easy, consistent installation
- Excellent for use with GC/MS since there’s no need to vent the MS when replacing the guard chip
- Not reusable, replace regularly for best results
- Best suited for ultra-clean applications, especially those using headspace or purge and trap sample introduction techniques
- Direct path from inlet to column
An Agilent Intuvo Guard Chip saves time and improves sample throughput by eliminating the need for column trimming. The power of a guard column is harnessed in an innovative metal microfluidic chip. It traps sample residue by providing almost one meter of inert flow path between the inlet liner and the column. With regular replacement, sample contamination never reaches the head of the Intuvo GC column - eliminating the need for column trimming.
The Intuvo Guard Chip is a disposable consumable. To prolong the lifetime of your Intuvo GC column, replace the guard chip at the same frequency as you would typically trim a conventional GC column, or whenever chromatographic performance begins to decline. Replacement is fast, consistent, and simple, minimising disruption to lab efficiency.
For some ultra-clean applications where a guard chip may not be needed, Intuvo Jumper Chips provide a short, direct flow path. Customers whose typical applications do not require column trimming may be able to use jumper chips. Most customer applications will utilise guard chips rather than jumper ships to ensure column longevity.
Achtung! Bitte beachten: Product only compatible with Intuvo 9000 GC System.
Technische Daten
- Bezeichnung:Guard chip, Intuvo, split/splitless inlet
- Seq:01