AuftragsverfolgungKontakt
Ersatzteile-Sets
Acquity SM performance maintenance kit. Injector valve cartridge (700005236) not included, rotor seal and pod repair kit included, ACQUITY UPLC Sample Mgr, 201000174, WATE
Catalog # CTSACTS-11141
Supplier:   SCIENCIX
Ersatzteile-Sets
Catalog # CTSACTS-11141
Supplier:  SCIENCIX
Ersatzteile-Sets
Ersatzteile-Sets
Acquity SM performance maintenance kit. Injector valve cartridge (700005236) not included, rotor seal and pod repair kit included, ACQUITY UPLC Sample Mgr, 201000174, WATE
Price
SFr. 1’980.00
Log in for availability
1 * 1 ST
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Specifications
  • Bezeichnung:
    Acquity SM performance maintenance kit. Injector valve cartridge (700005236) not included, rotor seal and pod repair kit included
  • Seq:
    17
  • Erstausrüster:
    WATE
  • Erstausrüster-Äquivalent:
    201000174
  • Modell:
    ACQUITY UPLC Sample Mgr