Ersatzteile-Sets
Acquity SM performance maintenance kit. Injector valve cartridge (700005236) not included, rotor seal and pod repair kit included, ACQUITY UPLC Sample Mgr, 201000174, WATE
Catalog # CTSACTS-11141
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Specifications
- Bezeichnung:Acquity SM performance maintenance kit. Injector valve cartridge (700005236) not included, rotor seal and pod repair kit included
- Seq:17
- Erstausrüster:WATE
- Erstausrüster-Äquivalent:201000174
- Modell:ACQUITY UPLC Sample Mgr