Racks for tubes
Blau, 25 (5×5), 5 ml
Katalognr. SIMPT450-25B
Anbieter: Simport Scientific
Direkt zu
Product Details & Documents
Made of polypropylene.
- Reusable, autoclavable and freezable (–90 °C)
- Wells are numbered for easier identification
- Good chemical resistance
Technische Daten
- Für Röhrchen:5 ml
- Anz. Bohrungen:25 (5×5)
- Farbe:Blau
- Seq:01